missing translation for 'onlineSavingsMsg'
Learn More

Monopropylheptylphthalate 6-Hydroxy-d4, TRC

Product Code. 30478388
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478388 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30478388 Supplier Toronto Research Chemicals Supplier No. TRCM56707710MG
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Monopropylheptylphthalate 6-Hydroxy-d4
CAS 1412411-11-0
Molecular Formula C18H22D4O5
Quantity 10 mg
Synonym OH-MiDP-d4, 1,2-Benzene-3,4,5,6-d4-dicarboxylic Acid 1-(6-Hydroxy-2-propylheptyl) Ester, OH-MPHP-d4
InChI Formula InChI=1S/C18H26O5/c1-3-7-14(9-6-8-13(2)19)12-23-18(22)16-11-5-4-10-15(16)17(20)21/h4-5,10-11,13-14,19H,3,6-9,12H2,1-2H3,(H,20,21)
SMILES OC(C1=C([2H])C([2H])=C([2H])C([2H])=C1C(OCC(CCC)CCCC(O)C)=O)=O
Molecular Weight (g/mol) 326.42
Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.