missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monostearyl Maleate, Mikromol™
Description
Monostearyl Maleate
Specifications
Specifications
| Chemical Name or Material | Monostearyl Maleate |
| CAS | 2424-62-6 |
| Analyte or Component Names | Monostearyl Maleate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H40 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Monostearyl Maleate |
| InChI Formula | InChI=1S/C22H40O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-20-26-22(25)19-18-21(23)24/h18-19H,2-17,20H2,1H3,(H,23,24)/b19-18- |
| SMILES | CCCCCCCCCCCCCCCCCCOC(=O)\C=C/C(=O)O |
| IUPAC Name | (Z)-4-octadecoxy-4-oxobut-2-enoic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?