missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Montmorillonite K 10, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Montmorillonite K 10 |
| CAS | 1318-93-0 |
| Molecular Formula | Al2H2O12Si4 |
| Quantity | 1 g |
| Synonym | 1090B, 1095B, AMS, AMS (mineral), Alabama Blue Clay, Albagen 4439, Amcol A |
| InChI Formula | InChI=1S/2Al.4O2Si.H2O.3O/c;;4*1-3-2;;;;/h;;;;;;1H2;;;/q2*+3;;;;;;3*-2 |
| SMILES | O.[O-2].[O-2].[O-2].O=[Si]=O.O=[Si]=O.O=[Si]=O.O=[Si]=O.[Al+3].[Al+3] |
| Molecular Weight (g/mol) | 360.31 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction