missing translation for 'onlineSavingsMsg'
Learn More

Morazone, TRC

Product Code. 30135804
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135804 100 mg 100mg
30003418 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135804 Supplier Toronto Research Chemicals Supplier No. TRCM630800100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6536-18-1
Molecular Formula C23 H27 N3 O2
Molecular Weight (g/mol) 377.48
Synonym 3H-Pyrazol-3-one, 1,2-dihydro-1,5-dimethyl-4-[(3-methyl-2-phenyl-4-morpholinyl)methyl]-2-phenyl-, Antipyrine, 4-[(3-methyl-2-phenylmorpholino)methyl]- (6CI,8CI), 4-[(3-Methyl-2-phenylmorpholino)methyl]antipyrine, Morazone, NSC 292156, Novartrina, Orsimon, R 445, Tarugan
IUPAC Name 1,5-dimethyl-4-[(3-methyl-2-phenylmorpholin-4-yl)methyl]-2-phenylpyrazol-3-one
SMILES CC1C(OCCN1CC2=C(C)N(C)N(C2=O)c3ccccc3)c4ccccc4

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C23H27N3O2/c1-17-21(23(27)26(24(17)3)20-12-8-5-9-13-20)16-25-14-15-28-22(18(25)2)19-10-6-4-7-11-19/h4-13,18,22H,14-16H2,1-3H3
Formula Weight 377.2103
Chemical Name or Material Morazone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.