missing translation for 'onlineSavingsMsg'
Learn More

Morin Hydrate, TRC

Product Code. 30481019
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30481019 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30481019 Supplier Toronto Research Chemicals Supplier No. TRCM63081310G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Morin Hydrate
CAS 654055-01-3
Molecular Formula C15 H10 O7 . H2 O
Quantity 10 g
InChI Formula InChI=1S/C15H10O7.H2O/c16-6-1-2-8(9(18)3-6)15-14(21)13(20)12-10(19)4-7(17)5-11(12)22-15;/h1-5,16-19,21H;1H2
SMILES O.OC1=C(Oc2cc(O)cc(O)c2C1=O)c3ccc(O)cc3O
IUPAC Name 2-(2,4-dihydroxyphenyl)-3,5,7-trihydroxychromen-4-one;hydrate
Molecular Weight (g/mol) 320.251
Formula Weight 320.053
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.