missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Morphine Monoclonal Antibody (2H2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549675
This item is not returnable.
View return policy
Description
Antibody tested in indirect ELISA.
Morphine Base is an opiate alkaloid isolated from the plant Papaver somniferum and produced synthetically. Morphine binds to and activates specific opiate receptors (delta, mu and kappa), each of which are involved in controlling different brain functions. In the central nervous and gastrointestinal systems, this agent exhibits widespread effects including analgesia, anxiolysis, euphoria, sedation, respiratory depression, and gastrointestinal system smooth muscle contraction. [National Center for Biotechnology Information. PubChem Compound Database; CID=5288826].
Specifications
| Morphine | |
| Monoclonal | |
| 0.91 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Mouse | |
| Protein G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry, Lateral Flow, SDS-Page | |
| 2H2 | |
| Unconjugated | |
| C17H19NO3; Morphia; Morphin; Morphinum; Morphium | |
| Conjugate of MOP hapten and KLH. | |
| 200 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction