missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Activin RIB/ALK-4 Antibody, R&D Systems™

Goat Polyclonal Antibody
209.00€ - 512.00€
Specifications
| Antigen | Activin RIB/ALK-4 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction |
| Applications | Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18724079
![]() |
R&D Systems
AF1477-SP |
25 μg |
209.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18078621
|
R&D Systems
AF1477 |
100 μg |
512.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Activin RIB/ALK-4 Polyclonal specifically detects Activin RIB/ALK-4 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction.Specifications
| Activin RIB/ALK-4 | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction | |
| Polyclonal | |
| Goat | |
| Mouse | |
| activin A receptor, type IB, Activin receptor type IB, Activin receptor-like kinase 4, ActivinRIB, ActRIB, ACVR1B, ACVRLK4ACTR-IB, ALK4, ALK-4, ALK4activin A receptor, type II-like kinase 4, EC 2.7.11, EC 2.7.11.30, Serine/threonine-protein kinase receptor R2, SKR2activin receptor type-1B | |
| ACVR1B | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction | |
| Unconjugated | |
| RUO | |
| Q61271 | |
| 91 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Activin RIB/ALK-4 Leu32-Glu126 Accession # Q61271 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse Activin RIB/ALK-4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant human (rh) Activin RIB is observed, 5% cross-reactivity with rhActivin R1A is observed and less than 1% cross-reactivity with rhActivin RIIA and rhActivin RIIB is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title