Learn More
CNOT7, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant CNOT7.
Brand: Abnova H00029883-A01.50uL
Description
The protein encoded by this gene binds to an anti-proliferative protein, B-cell translocation protein 1, which negatively regulates cell proliferation. Binding of the two proteins, which is driven by phosphorylation of the anti-proliferative protein, causes signaling events in cell division that lead to changes in cell proliferation associated with cell-cell contact. The protein has both mouse and yeast orthologs. Alternate splicing of this gene results in two transcript variants encoding different isoforms. [provided by RefSeq
Sequence: MKKIRQVIRKYNYVAMDTEFPGVVARPIGEFRSNADYQYQLLRCNVDLLKIIQLGLTFMNEQGEYPPGTSTWQFNFKFNLTEDMYAQDSIELLTTSGIQFSpecifications
| CNOT7 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant CNOT7. | |
| CNOT7 | |
| CAF1/hCAF-1 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_013354 | |
| CNOT7 | |
| CNOT7 (NP_037486, 1 a.a. to 100 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 29883 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only