Learn More
KRT18, Mouse, Biotin, Clone: C-04, Abnova™
Mouse monoclonal antibody raised against native KRT18.
Brand: Abnova MAB6504.100ug
Description
KRT18 encodes the type I intermediate filament chain keratin 18. Keratin 18, together with its filament partner keratin 8, are perhaps the most commonly found members of the intermediate filament gene family. They are expressed in single layer epithelial tissues of the body. Mutations in this gene have been linked to cryptogenic cirrhosis. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq
Specifications
| KRT18 | |
| Monoclonal | |
| Biotin | |
| The optimal working dilution should be determined by the end user. | |
| KRT18 | |
| KRT18 | |
| Native purified KRT18 from human epidermal carcinoma cell line (A-431). | |
| RUO | |
| 3875 | |
| Human, Mammalia | |
| Liquid |
| ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
| C-04 | |
| Mouse monoclonal antibody raised against native KRT18. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| CYK18/K18 | |
| Mouse | |
| 100 μg | |
| Primary | |
| This antibody reacts with Cytokeratin 18 (45 KDa), a member of intermediate filamentssubfamily represented in epithelial tissues. | |
| Store at 4°C. Do not freeze. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.