Learn More
PFKFB1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant PFKFB1.
Brand: Abnova H00005207-A01.50uL
Description
This gene encodes a member of the family of bifunctional 6-phosphofructo-2-kinase:fructose-2,6-biphosphatase enzymes. The enzyme forms a homodimer that catalyzes both the synthesis and degradation of fructose-2,6-biphosphate using independent catalytic domains. Fructose-2,6-biphosphate is an activator of the glycolysis pathway and an inhibitor of the gluconeogenesis pathway. Consequently, regulating fructose-2,6-biphosphate levels through the activity of this enzyme is thought to regulate glucose homeostasis. [provided by RefSeq
Sequence: MSPEMGELTQTRLQKIWIPHSSGSSRLQRRRGSSIPQFTNSPTMVIMVGLPARGKTYISTKLTRYLNWIGTPTKVFNLGQYRREAVSYKNYEFFLPDNMESpecifications
| PFKFB1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant PFKFB1. | |
| PFKFB1 | |
| F6PK/HL2K/MGC116715/MGC116717/PFRX | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_002625 | |
| PFKFB1 | |
| PFKFB1 (NP_002616, 1 a.a. ∽ 100 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 5207 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only