missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCL1/I-309/TCA-3 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF845-SP
This item is not returnable.
View return policy
Description
CCL1/I-309/TCA-3 Polyclonal specifically detects CCL1/I-309/TCA-3 in Mouse samples. It is validated for Western Blot, Neutralization.
Specifications
| CCL1/I-309/TCA-3 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C-C motif chemokine 1, chemokine (C-C motif) ligand 1, I-309, inflammatory cytokine I-309, P500, SCYA1, SCYA1Small-inducible cytokine A1, SCYA2, SISe, small inducible cytokine A1 (I-309, homologous to mouse Tca-3), T lymphocyte-secreted protein I-309, TCA3, TCA-3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6346 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.3-1.5 ug/mL | |
| P10146 | |
| CCL1 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse CCL1/I-309/TCA-3 Lys24-Cys92 Accession # P10146 | |
| 25 μg | |
| Primary | |
| Detects mouse CCL1 in direct ELISAs and Western blots. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction