missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD300a/LMIR1 PE-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems FAB1186P
This item is not returnable.
View return policy
Description
CD300a/LMIR1 Monoclonal specifically detects CD300a/LMIR1 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CD300a/LMIR1 | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD300a, CD300a antigenCMRF35H9, CD300a molecule, CLM8, CLM-8, CMRF-35H, CMRF35-H, CMRF35H leukocyte immunoglobulin-like receptor, CMRF35-H9, CMRF-35-H9CD300 antigen-like family member A, CMRF35HIRp60, CMRF35-like molecule 8, IgSF12, IGSF12NK inhibitory receptor, Immunoglobulin superfamily member 12, Inhibitory receptor protein 60, IRC1, IRC1/IRC2, IRC2, IRp60, leukocyte membrane antigen, LMIR1, MAIR-I, Mcipr1, NKRL, Pigr4 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 11314 | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 172224 | |
| Flow Cytometry 10 uL/10^6 cells | |
| BAC80268 | |
| CD300A | |
| Mouse myeloma cell line NS0-derived recombinant mouse CD300a/LMIR1 Leu28-Arg183 Accession # BAC80268 | |
| 100 Tests | |
| Primary | |
| Detects mouse CD300a/LMIR1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human LMIR1, 2, 3, 4, 5, 6, recombinant mouse LMIR3, 4, or 5 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction