missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD47 Antibody, R&D Systems™

Rat Monoclonal Antibody
196.00€ - 447.00€
Specifications
| Antigen | CD47 |
|---|---|
| Clone | 974214 |
| Dilution | Flow Cytometry 0.25 μg/106 cells, CyTOF-ready |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18752767
![]() |
R&D Systems
MAB1866-SP |
25 μg |
196.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18763157
![]() |
R&D Systems
MAB1866-100 |
100 μg |
447.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD47 Monoclonal specifically detects CD47 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| CD47 | |
| Flow Cytometry 0.25 μg/106 cells, CyTOF-ready | |
| Monoclonal | |
| Rat | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| antigen identified by monoclonal 1D8, Antigenic surface determinant protein OA3, CD47, CD47 antigen, CD47 antigen (Rh-related antigen, integrin-associated signal transducer), CD47 glycoprotein, CD47 molecule, IAP, IAPintegrin associated protein, Integrin-associated protein, leukocyte surface antigen CD47, MER6, MER6integrin-associated signal transducer, OA3, Protein MER6, Rh-related antigen | |
| Mouse myeloma cell line, NS0-derived mouse CD47 protein, Gln19-Pro158, Accession # NP_034711 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse CD47 in direct ELISAs. |
| 974214 | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Mouse | |
| NP_034711 | |
| 961 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title