missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Coagulation Factor VII Antibody, R&D Systems™

Rat Monoclonal Antibody
176.00€ - 378.00€
Specifications
| Antigen | Coagulation Factor VII |
|---|---|
| Clone | 406722 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18774578
![]() |
R&D Systems
MAB3305-SP |
25 μg |
176.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18192521
![]() |
R&D Systems
MAB3305 |
100 μg |
378.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Coagulation Factor VII Monoclonal specifically detects Coagulation Factor VII in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Coagulation Factor VII | |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rat | |
| Mouse | |
| P70375 | |
| 2155 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Coagulation Factor VII Ala42-Leu446 Accession # P70375 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse Coagulation Factor VII in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) FGFA, rhFactor VII, rhFactor X, rhFactor XI, rhPROC, rhThrombin, rhUPA1, rhKLK-1, -2, -3, -4, -5, -7, -8, -9, -10, -11, -12, -13, -14, -15, or -B1 is observed. |
| 406722 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| coagulation factor VII, coagulation factor VII (serum prothrombin conversion accelerator), EC 3.4.21, EC 3.4.21.21, Eptacog alfa, F7, FVII coagulation protein, Proconvertin, Serum prothrombin conversion accelerator, SPCA | |
| F7 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title