missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CXCR6 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB2145N-025
This item is not returnable.
View return policy
Description
CXCR6 Monoclonal specifically detects CXCR6 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CXCR6 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Q9EQ16 | |
| CXCR6 | |
| CHO Chinese hamster ovary cell line transfected with mouse CXCR6 Met1-Leu351 Accession # Q9EQ16 | |
| 25 Tests | |
| 10663 | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 221002 | |
| Flow Cytometry 5 uL/10^6 cells | |
| Bonzo, BONZOG-protein coupled receptor bonzo, CD186, CD186 antigen, CDw186, chemokine (C-X-C motif) receptor 6, C-X-C chemokine receptor type 6, CXCR6, CXC-R6, CXCR-6, G protein-coupled receptor, STRL33, STRL33G-protein coupled receptor STRL33, TYMSTRCD186 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Stains mouse CXCR6-transfected cells but not irrelevant transfectants. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction