missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse DCC (NP_031857) VersaClone cDNA
Description
- Also known as: Igdcc1; C030036D22Rik
- DCC is a transmembrane protein that is a member of the immunoglobulin superfamily.
- DCC is expressed on axons during development where its binding to Netrin-1 regulates axon migration and myelination. It inhibits apoptosis when bound by Netrin-1 and can induce apoptosis in the absence of ligand.
- DCC also functions as an adhesion molecule and a tumor suppressor on intestinal epithelial cells.
- Deficiencies in DCC function are associated with the development and metastasis of many tumors.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mDcc |
| Accession Number | NP_031857 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 4357 bp |
| Gene ID (Entrez) | 1630 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Product Line | R&D Systems |
| Quantity | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?