Learn More
R&D Systems™ Mouse EDA2R/TNFRSF27/XEDAR (NP_780749) VersaClone cDNA
Product Code. 18727769
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18727769 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18727769 Supplier R&D Systems™ Supplier No. RDC0937
Ectodysplasin A2 receptor
The Mouse EDA2R/TNFRSF27/XEDAR (NP_780749) VersaClone cDNA clone contains the complete ORF for EDA2R/TNFRSF27/XEDAR, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The EDA2R/TNFRSF27/XEDAR insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 10 μg
For Reasearch Use Only; Not for Use in Humans