missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse G6b (NP_001028393) VersaClone cDNA

Product Code. 18700769
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18700769 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18700769 Supplier R&D Systems™ Supplier No. RDC2561
  • ONLINE EXCLUSIVE PRICE
    328.00€
    309.75€ / 10µg
    Save 18.25€
    5% Off
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.

  • Mpig6b megakaryocyte and platelet inhibitory receptor G6b [ Mus musculus (house mouse) ]
  • Also known as: G6b; Ng31; C6orf25; AU023871
  • The G6b/C6orf25 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
  • G6B is a member of the immunoglobulin (Ig) superfamily and is located in the major histocompatibility complex (MHC) class III region.
  • It is a glycosylated, plasma membrane-bound cell surface receptor, but soluble isoforms have been found in the endoplasmic reticulum and Golgi before being secreted.
  • It is expressed mainly on the platelet lineage. Crosslinking of G6b inhibits agonist-stimulated platelet aggregation and activation.
  • Heparin interacts with the G6b extracellular domain with high-affinity.
  • For reasearch use only. Not for use in humans.
TRUSTED_SUSTAINABILITY

Specifications

Gene mMpig6b
Accession Number NP_001028393
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Insert Type 607 bp
Gene ID (Entrez) 80739
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Mouse
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.