missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Galectin-2 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB6667-SP
This item is not returnable.
View return policy
Description
Galectin-2 Monoclonal specifically detects Galectin-2 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Galectin-2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| Q9CQW5 | |
| LGALS2 | |
| E. coli-derived recombinant mouse Galectin-2 Ser2-Glu130 Accession # Q9CQW5 | |
| 25 μg | |
| Primary | |
| Detects mouse Galectin-2 in direct ELISAs and Western blots. In Western blots, approximately 5% cross-reactivity with recombinant human (rh) Galectin-2 is observed and no cross-reactivity with rhGalectin-4, -8, -10, recombinant mouse Galectin-1, -3, -7, or -9 is observed. | |
| Mouse | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 747307 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Beta-galactoside-binding lectin L-14-II, Gal-2, GAL2, galectin 2, galectin-2, Galectin2, HL14gal-2, Lactose-binding lectin 2, lectin, galactoside-binding, soluble, 2, LGALS2, MGC75071, S-Lac lectin 2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3957 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction