missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse GPVI Alexa Fluor 750-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB6758S-100UG
This item is not returnable.
View return policy
Description
GPVI Monoclonal specifically detects GPVI in Mouse samples. It is validated for Flow Cytometry.
Specifications
| GPVI | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P0C191 | |
| GP6 | |
| Mouse myeloma cell line NS0-derived recombinant mouse GPVI Gly24-Lys265 Accession # P0C191 | |
| RUO | |
| 51206 | |
| Mouse | |
| IgG1 |
| Flow Cytometry | |
| 784808 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Glycoprotein 6, glycoprotein VI (platelet), GP6, GPIV, GPVIplatelet collagen receptor, MGC138168, platelet glycoprotein VI | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse GPVI in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human GPVI is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction