missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Melanocortin-3 R/MC3R Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB3737-SP
This item is not returnable.
View return policy
Description
Melanocortin-3 R/MC3R Monoclonal specifically detects Melanocortin-3 R/MC3R in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| Melanocortin-3 R/MC3R | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| BMIQ9, MC3, MC3R, MC3-R, melanocortin 3 receptor, melanocortin receptor 3, Melanocortin3R, Melanocortin-3R, OB20, Obesity quantitative trait locus, OQTL | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4159 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Flow Cytometry, CyTOF | |
| 345404 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MC3R | |
| Y3 rat myeloid cell line transfeced with mouse Melanocortin 3R/MC3R | |
| 25 μg | |
| Primary | |
| Detects Melanocortin 3R/MC3R in direct ELISAs and Western blots. Stains mouse Melanocortin 3R/MC3R-transfected cells but not irrelevant transfectants. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction