Learn More
Mouse IFN-beta Antibody, R&D Systems™
GREENER_CHOICE
Rabbit Monoclonal Antibody
Brand:  R&D Systems MAB8234-SP
 View more versions of this product
Product Code. 18765199
  • 209.00€ / 25µg
Description
IFN-beta Monoclonal specifically detects IFN-beta in Mouse samples. It is validated for Neutralization.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IFN-beta
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956
Human embryonic kidney cell line HEK293-derived mouse IFN-beta Ile22-Asn182 Accession # P01575
25 μg
Primary
Detects mouse IFN-beta in direct ELISAs.
Mouse
IgG
Neutralization
1176D
Neutralization 0.25-1.25 ug/mL
P01575
Rabbit
Protein A or G purified from cell culture supernatant
RUO
3456
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.