missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IFN-gamma Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB485R-SP
This item is not returnable.
View return policy
Description
IFN-gamma Monoclonal specifically detects IFN-gamma in Mouse samples. It is validated for Neutralization, Intracellular Staining by Flow Cytometry.
Specifications
| IFN-gamma | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Ifng | |
| E. coli-derived recombinant mouse IFN-gamma | |
| 25 μg | |
| Primary | |
| Detects mouse IFN-gamma in Western blots. In Western blots, this antibody does not cross-react with recombinant human (rh) IFN-gamma , rrIFN-gamma , rpIFN-gamma , rrmIFN-gamma , rfeIFN-gamma , or rcaIFN-gamma . | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Neutralization, Flow Cytometry | |
| 37895R | |
| Neutralization 0.075-0.3 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells | |
| IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
| Rat | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3458 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction