missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC76 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32177AF647
This item is not returnable.
View return policy
Description
CCDC76 Polyclonal antibody specifically detects CCDC76 in Human, Mouse samples. It is validated for Western Blot
Specifications
| CCDC76 | |
| Polyclonal | |
| Western Blot | |
| coiled-coil domain containing 76, Coiled-coil domain-containing protein 76, EC 2.1.1.-, FLJ10287, FLJ11219, tRNA [Gm4] methyltransferase, tRNA guanosine-2'-O-methyltransferase TRM13 homolog | |
| The immunogen this antibody was made to, maps to a region between residue 25 to 75 of human Coiled-Coil Domain Containing 76 using the numbering given in entry NP_061956.2 (GeneID 54482). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 54482 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction