missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IgLON5 (NP_001157990.1) VersaClone cDNA

Product Code. 18710687
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18710687 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18710687 Supplier R&D Systems™ Supplier No. RDC3483
  • ONLINE EXCLUSIVE PRICE
    328.00€
    309.75€ / 10µg
    Save 18.25€
    5% Off
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation.

Inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.
TRUSTED_SUSTAINABILITY

Specifications

Gene IGLON5
Accession Number NP_001157990.1
Concentration 10 μg at 0.2 μg/ μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Insert Type 1024 bp
Gene ID (Entrez) 402665
Gene Alias hCG1651476, Ig-like domain-containing protein ENSP00000270642, IgLON family member 5
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Mouse
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.