missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-12/IL-23 p40 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB499-500
This item is not returnable.
View return policy
Description
IL-12/IL-23 p40 Monoclonal specifically detects IL-12/IL-23 p40 in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
| IL-12/IL-23 p40 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Il12b | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL-12 p40 | |
| 500 μg | |
| Primary | |
| Detects mouse IL-12 p40 in ELISAs. Polymorphisms exist in the mouse IL-12 p40 sequence. This antibody recognizes an allelic epitope found in the p40 subunit of several mouse strains including BALB/c, C57BL/6, CBA, C3H, DBA/1, NOR, I29/J, and A.SW. This antibody does not recognize the allelic variant that occurs in strains AKR, DBA/2, NOD, NZB, NZC, NZO, NZW, or SJL (1). In ELISAs, this antibody shows 100% cross-reactivity with recombinant mouse (rm) IL-23, 30% cross-reactivity with rmIL-12 p70, and no cross-reactivity with recombinant human (rh) IL-12 p70 or rhIL-12 p40. | |
| Mouse | |
| Supernatant |
| ELISA | |
| 30517 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL12 p40, IL-12 subunit p40, IL12B, IL-12B, IL-12BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2, NKSF2IL12, subunit p40 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 16160 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction