missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-22 R alpha 1 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB42941V-100UG
This item is not returnable.
View return policy
Description
IL-22R alpha 1 Monoclonal specifically detects IL-22R alpha 1 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| IL-22R alpha 1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q80XZ4 | |
| IL22RA1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-22 R alpha 1 Thr18-Ala228 Accession # Q80XZ4 | |
| RUO | |
| 58985 | |
| Mouse | |
| IgG2a |
| Flow Cytometry | |
| 496514 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CRF2-9, CRF2-9interleukin 22 receptor, Cytokine receptor class-II member 9, Cytokine receptor family 2 member 9, IL-22 receptor subunit alpha-1, IL22R, IL22R alpha 1, IL22R1, IL22RA1, IL-22RA1, IL-22R-alpha-1, IL-TIF-R1, interleukin 22 receptor, alpha 1, interleukin-22 receptor subunit alpha-1, zcytoR11 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse IL-22 R alpha 1 in direct ELISAs. In direct ELISAs, 25% cross-reactivity with recombinant human (rh) IL-22 R alpha 1 is observed and no cross-reactivity with rhIL-20 R alpha , recombinant mouse (rm) IL-20 R alpha , rhIL-22BP, or rmIL-22BP is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction