missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-23R Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Brand: R&D Systems AF1686
This item is not returnable.
View return policy
Description
IL-23R Polyclonal specifically detects IL-23R in Mouse samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.
Specifications
| IL-23R | |
| Polyclonal | |
| Unconjugated | |
| Q8K4B4 | |
| IL23R | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-23 R Gly24-Asp372 Accession # Q8K4B4 | |
| 100 μg | |
| Primary | |
| Detects mouse IL-23 R in direct ELISAs and Western blots. In these formats, approximately 15% cross-reactivity with recombinant human IL-23 R is observed. | |
| Mouse | |
| IgG |
| Western Blot, Blockade of Receptor-ligand Interaction | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction | |
| IBD17, IL-23 R, IL-23 receptor, IL23R, IL-23R, interleukin 23 receptor, interleukin-23 receptor | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 149233 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction