missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse MARCO (NP_034896.1) VersaClone cDNA

Product Code. 18793547
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18793547 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18793547 Supplier R&D Systems™ Supplier No. RDC3425
  • ONLINE EXCLUSIVE PRICE
    328.00€
    309.75€ / 10µg
    Save 18.25€
    5% Off
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Contains gene inserts that are fully sequenced and produced with high quality

TRUSTED_SUSTAINABILITY

Specifications

Gene MARCO
Accession Number NP_034896.1
Concentration 10 μg (at 0.2 μg/μL)
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Insert Type 1570 bp
Gene ID (Entrez) 8685
Gene Alias macrophage receptor with collagenous structureSCARA2Scavenger receptor class A member 2, member 2, SCARA2
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Mouse
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.