missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD97 v2 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB3734T-100UG
This item is not returnable.
View return policy
Description
CD97 Isoform 1 Monoclonal specifically detects CD97 Isoform 1 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CD97 Isoform 1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| AAH06676 | |
| CD97 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse CD97v2 Gln24-His384 Accession # AAH06676 | |
| 100 μg | |
| Primary | |
| In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse (rm) CD97 and no cross-reactivity with recombinant human (rh) CD96v2, rhCD97, or rmCD96 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| Flow Cytometry | |
| 587702 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD97, TM7LN1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 976 | |
| Mouse | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction