missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse OC-STAMP (NP_083297) VersaClone cDNA

Product Code. 18761009
Change view
Click to view available options
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18761009 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18761009 Supplier R&D Systems™ Supplier No. RDC2670

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.

  • Also known as: OC-STAMP; 4833422F24Rik
  • OCSTAMP is very strongly upregulated during osteoclast differentiation.
  • It is a multipass transmembrane protein that is highly conserved in mammals and has little homology to other proteins.
  • OCSTAMP cooperates with DCSTAMP in modulating cell-cell fusion in both osteoclasts and foreign body giant cells (FBGCs).
  • It is involved in osteoclast bone resorption and promotes osteoclast differentiation.
  • OCSTAMP may play a role in multinucleated osteoclast maturation.
  • The OC-STAMP insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
  • For reasearch use only. Not for use in humans.
TRUSTED_SUSTAINABILITY

Specifications

Gene mOcstamp
Accession Number NP_083297
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Insert Type 1510 bp
Gene ID (Entrez) 128506
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Mouse
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.