missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse OCIL/CLEC2d (NP_444339) VersaClone cDNA

Product Code. 18770798
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18770798 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18770798 Supplier R&D Systems™ Supplier No. RDC2548
  • ONLINE EXCLUSIVE PRICE
    328.00€
    309.75€ / 10µg
    Save 18.25€
    5% Off
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.

  • Clec2d C-type lectin domain family 2, member d [ Mus musculus (house mouse) ]
  • Also known as: Clrb; Ocil; Clr-b
  • The OCIL/CLEC2d insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
  • CLEC2D is a type II transmembrane protein belonging to the natural killer cell receptor C-type lectin family.
  • It is found on hematopoietic cells, osteoblasts, and chondrocytes.
  • CLEC2D inhibits osteoclast formation and contains a transmembrane domain near the Nterminus as well as the C-type lectin-like extracellular domain.
  • For reasearch use only. Not for use in humans.
TRUSTED_SUSTAINABILITY

Specifications

Gene mClec2d
Accession Number NP_444339
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Insert Type 637 bp
Gene ID (Entrez) 29121
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Mouse
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.