missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Mouse Patched 1/PTCH Antibody, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals™ MAB11487-SP
This item is not returnable.
View return policy
Description
Patched 1/PTCH Monoclonal antibody specifically detects Patched 1/PTCH in Mouse samples. It is validated for Immunohistochemistry,Immunocytochemistry
Specifications
| Patched 1/PTCH | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. See Certificate of Analysis for details. *Small pack size (-SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| BCNS, BCNSFLJ42602, FLJ26746, HPE7, mes, NBCCS, patched (Drosophila) homolog, patched 1, patched homolog (Drosophila), patched homolog 1, patched homolog 1 (Drosophila), protein patched homolog 1, PTC, PTC1, PTCH, PTCH protein +12b, PTCH protein +4', PTCH protein -10, PTCH1, PTCH11 | |
| E. coli -derived recombinant mouse Patched 1/PTCH, Glu108-Asp422, Accession # Q61115 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Use a manual defrost freezer and avoid repeated freeze/thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. | |
| IgG2a |
| Immunohistochemistry, Immunocytochemistry | |
| 413218 | |
| Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL | |
| Q61115 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5727 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction