Learn More
Mouse PD-L1/B7-H1 Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Brand:  R&D Systems MAB11630-SP
 View more versions of this product
Product Code. 30306072
  • 247.00€ / 25µg
Description
PD-L1 Monoclonal antibody specifically detects PD-L1 in Mouse samples. It is validated for Immunohistochemistry,Multiplex Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PD-L1
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Avelumab, B7-H, B7H1, B7-H1, B7H1PDCD1L1, CD274, CD274 antigenMGC142294, CD274 molecule, PDCD1L1, PDCD1LG1, PDCD1LG1MGC142296, PDL1, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1
Mouse myeloma cell line, NS0 derived mouse PD-L1/B7-H1, Phe19-His239, Accession # Q9EP73
25 μg
Primary
Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Immunohistochemistry, Multiplex Immunofluorescence
2096A
Immunohistochemistry 0.05-10 μg/mL, Multiplex Immunofluorescence 5 μg/mL
Q9EP73
Rabbit
Protein A or G purified from cell culture supernatant
RUO
29126