missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Podocalyxin Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB1556
This item is not returnable.
View return policy
Description
Podocalyxin Like Monoclonal specifically detects Podocalyxin Like in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready.
Specifications
| Podocalyxin Like | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 μg/mL, Flow Cytometry 2.5 μg/10^6 cells, Immunohistochemistry 1-25 μg/mL, Immunocytochemistry 8-25 μg/mL, CyTOF-ready | |
| Q9R0M4 | |
| PODXL | |
| Mouse myeloma cell line NS0-derived recombinant mouse Podocalyxin Ser21-Arg402 Accession # Q9R0M4 | |
| 100 μg | |
| Primary | |
| Detects mouse Podocalyxin in direct ELISAs and Western blots. | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF | |
| 192703 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| GCTM, Gp200MGC138240, PCLP, PCLP1, PCLP-1, PCLPGCTM-2 antigen, PCpodocalyxin, POD XL, podocalyxin-like, Podocalyxin-like protein 1, PODXL | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5420 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction