missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
R&D Systems™ Mouse RAGE (NP_031451) VersaClone cDNA
Beskrivning
The Mouse RAGE (NP_031451) VersaClone cDNA clone contains the complete ORF for AGER, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The AGER insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Beställningsinfo
Ships at ambient temperature.
Specifikationer
Specifikationer
| Product Type | cDNA |
| Gene | mRAGE |
| Accession Number | NP_031451 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10mM Tris-Cl, pH 8.5 |
| Insert Type | 1221bp |
| Gene ID (Entrez) | 5891 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Quantity | 10 μg |
For Research Use Only. Not for use in Humans.
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?