missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse/Rat Leptin Quantikine ELISA Kit
Specifications
Specifications
| Assay Range | 31.30 to 2000 pg/mL |
| Assay Sensitivity | 5.56 pg/mL |
| Sample Type | Cell Culture Supernatant, Mouse Plasma (EDTA), Mouse Plasma (Heparin), Mouse Serum, Rat Plasma (EDTA), Rat Plasma (Heparin), Rat Serum, Tissue Homogenates |
| Specificity | Natural and Recombinant Mouse and Rat Leptin |
| Target Species | Mouse, Rat |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3952 |
| Quantity | 1 Pk. (6 Plates) |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Synonym | FLJ94114, LEP, leptin, leptin (murine obesity homolog), leptin (obesity homolog, mouse), OB, Obese protein, obese, mouse, homolog of, Obesity factor, OBOBS |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?