missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Mouse Recombinant Flt3/Flk-2 Ligand
Description
- Flt3/Flk-2 (Fms-like tyrosine kinase 3/fetal liver kinase-2) ligand is a hematopoietic cytokine that plays an important role as a co-stimulatory factor in the proliferation, differentiation, and survival of hematopoietic stem and progenitor cells and the development of the immune system
- Flt3/Flk-2 ligand, together with stem cell factor and thrombopoietin, is commonly used to promote expansion of primitive hematopoietic cells in culture
- In combination with myeloid cytokines such as GM-CSF, G-CSF, or M-CSF, Flt3/Flk-2 ligand enhances the growth and numbers of clonogenic myeloid progenitor cells
- In synergy with IL-3, IL-4, IL-7, IL-11, IL-12, IL-15, GM-CSF, and TNF-α, Flt3/Flk-2 ligand regulates the development of various lymphoid progenitor cells, including dendritic cell, B cell, T cell, and NK cell progenitors
- In contrast, Flt3/Flk-2 ligand has no significant effect on erythropoiesis or megakaryopoiesis
Specifications
Specifications
| Name | FL, FLT3L, Flt3-L, Flt3 Ligand, Fms-like tyrosine kinase 3 ligand, Fms-like tyrosine kinase 3/fetal liver kinase-2 |
| Quantity | 10 μg |
| Cell Type | Dendritic Cells; Hematopoietic Stem and Progenitor Cells; NK Cells |
| Purity or Quality Grade | ≥95% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?