missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Mouse Recombinant Flt3/Flk-2 Ligand

Product Code. 17149341
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
10µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17149341 100 μg 100µg
17159341 10 μg 10µg
17169341 1000 μg 1000µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17149341 Supplier STEMCELL Technologies Supplier No. 78011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fms-like tyrosine kinase 3/fetal liver kinase-2

  • Flt3/Flk-2 (Fms-like tyrosine kinase 3/fetal liver kinase-2) ligand is a hematopoietic cytokine that plays an important role as a co-stimulatory factor in the proliferation, differentiation, and survival of hematopoietic stem and progenitor cells and the development of the immune system
  • Flt3/Flk-2 ligand, together with stem cell factor and thrombopoietin, is commonly used to promote expansion of primitive hematopoietic cells in culture
  • In combination with myeloid cytokines such as GM-CSF, G-CSF, or M-CSF, Flt3/Flk-2 ligand enhances the growth and numbers of clonogenic myeloid progenitor cells
  • In synergy with IL-3, IL-4, IL-7, IL-11, IL-12, IL-15, GM-CSF, and TNF-α, Flt3/Flk-2 ligand regulates the development of various lymphoid progenitor cells, including dendritic cell, B cell, T cell, and NK cell progenitors
  • In contrast, Flt3/Flk-2 ligand has no significant effect on erythropoiesis or megakaryopoiesis

Specifications

Name FL, FLT3L, Flt3-L, Flt3 Ligand, Fms-like tyrosine kinase 3 ligand, Fms-like tyrosine kinase 3/fetal liver kinase-2
Quantity 100 μg
Cell Type Dendritic Cells; Hematopoietic Stem and Progenitor Cells; NK Cells
Purity or Quality Grade ≥95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.