missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Resistin Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB1069-SP
This item is not returnable.
View return policy
Description
Resistin Monoclonal specifically detects Resistin in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications
| Resistin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q99P87 | |
| RETN | |
| E. coli-derived recombinant mouse Resistin Ser21-Ser114 Accession # Q99P87 | |
| 25 μg | |
| Primary | |
| Detects mouse Resistin in ELISAs and Western blots. Does not cross-react with recombinant mouse RELM alpha . | |
| Mouse | |
| Purified |
| Western Blot, ELISA | |
| 175322 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Adipose tissue-specific secretory factor, ADSF, ADSFMGC126609, C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich proteinprecursor 1, C/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein, Cysteine-rich secreted protein A12-alpha-like 2, Cysteine-rich secreted protein FIZZ3, FIZZ3, FIZZ3MGC126603, found in inflammatory zone 3, HXCP1, resistin, RETN, RETN1, RSTNXCP1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 56729 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction