missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse TfR (NP_035768.1) VersaClone cDNA

Product Code. 18723557
Change view
Click to view available options
:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18723557 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18723557 Supplier R&D Systems™ Supplier No. RDC3149

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Contains gene inserts that are fully sequenced and produced with high quality

TRUSTED_SUSTAINABILITY

Specifications

Gene TfR (Transferrin R)
Accession Number NP_035768.1
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Insert Type 2305 bp
Gene ID (Entrez) 7037
Gene Alias CD71, CD71 antigen, CD71TRFR, p90, T9, TfR, TfR (TransferrinR), TFR1, TFRC, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Mouse
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.