missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse TROP-2 (NP_064431.2) VersaClone cDNA

Product Code. 18726878
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18726878 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18726878 Supplier R&D Systems™ Supplier No. RDC3068
  • ONLINE EXCLUSIVE PRICE
    341.00€
    322.35€ / 10µg
    Save 18.65€
    5% Off
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Contains gene inserts that are fully sequenced and produced with high quality

TRUSTED_SUSTAINABILITY

Specifications

Gene TROP-2
Accession Number NP_064431.2
Gene ID (Entrez) 4070
Gene Alias Cell surface glycoprotein Trop-2, EGP1, EGP-1, epithelial glycoprotein-1, GA733-1, GA733-1 GP50, gastrointestinal tumor-associated antigen GA7331, gp50, M1S1cell surface glycoprotein TROP2, Membrane component chromosome 1 surface marker 1, membrane component, chromosome 1, surface marker 1, Pancreatic carcinoma marker protein GA733-1, pancreatic carcinoma marker protein GA7331,40 kD glycoprotein, identified by monoclonal GA733, T16, TACSTD2, TROP2, TROP2GA7331, tumor-associated calcium signal transducer 2
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Mouse
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.