missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TROY/TNFRSF19 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF723
This item is not returnable.
View return policy
Description
TROY/TNFRSF19 Polyclonal specifically detects TROY/TNFRSF19 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, ELISA Capture (Matched Antibody Pair).
Specifications
| TROY/TNFRSF19 | |
| Polyclonal | |
| Unconjugated | |
| Q9JLL3 | |
| TNFRSF19 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse TROY/TNFRSF19 Glu30-Leu170 Accession # Q9JLL3 | |
| 100 μg | |
| Primary | |
| Detects mouse TROY/TNFRSF19 in ELISAs and Western blots. In sandwich immunoassays, less than 2% cross-reactivity with recombinant human TAJ is observed and less than 0.2% cross-reactivity with recombinant mouse (rm) EDAR, rmCD27, rmFas, rmRANK, rmTNF RI, and rmTNF RII is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| TAJ, TAJ-alpha, TAJToxicity and JNK inducer, TNFRSF19, TRADE, TROYtumor necrosis factor receptor superfamily member 19, tumor necrosis factor receptor superfamily, member 19 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 55504 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction