missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TWEAK/TNFSF12 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF1237
This item is not returnable.
View return policy
Description
TWEAK/TNFSF12 Polyclonal specifically detects TWEAK/TNFSF12 in Mouse samples. It is validated for Western Blot, Neutralization.
Specifications
| TWEAK/TNFSF12 | |
| Polyclonal | |
| Unconjugated | |
| O54907 | |
| Tnfsf12 | |
| E. coli-derived recombinant mouse TWEAK/TNFSF12 Arg105-His249 Accession # O54907 | |
| 100 μg | |
| Primary | |
| Detects TWEAK/TNFSF12 in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant human TWEAK is observed and less than 1% cross-reactivity with recombinant mouse (rm) BAFF, rmFas Ligand, rmOX40 Ligand, rmTNF-alpha , and rmTRAIL is observed. | |
| Mouse | |
| IgG |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 4-16 ug/mL | |
| APO3 ligand, APO3LMGC20669, DR3LGTWEAKAPO3/DR3 ligand, MGC129581, TNF-related weak inducer of apoptosis, TNFSF12, tumor necrosis factor (ligand) superfamily, member 12, tumor necrosis factor ligand superfamily member 12 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8742 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction