missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse WISP-1/CCN4 Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF1680-SP
This item is not returnable.
View return policy
Description
WISP-1/CCN4 Polyclonal specifically detects WISP-1/CCN4 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| WISP-1/CCN4 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CCN family member 4, CCN4, CCN4WISP1tc, WISP1, WISP-1, WISP1c, WISP1i, WNT1 induced secreted protein 1, Wnt-1 inducible signaling pathway protein 1, WNT1 inducible signaling pathway protein 1, wnt-1 signaling pathway protein 1, Wnt-1-induced secreted protein, WNT1-inducible-signaling pathway protein 1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 8840 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| O54775 | |
| WISP1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse WISP-1/CCN4 Thr23-Asn367 Accession # O54775 | |
| 25 μg | |
| Primary | |
| Detects mouse WISP-1/CCN4 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human (rh) WISP-1 is observed and less than 1% cross-reactivity with rhCTGF and recombinant mouse NOV is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction