missing translation for 'onlineSavingsMsg'
Learn More

Moxalactam sodium salt

Product Code. 15750899 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15750899 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code. 15750899 Supplier Thermo Scientific Acros Supplier No. 460410010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

CAS: 64953-12-4 | C20H18N6Na2O9S | 564.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 64953-12-4
Molecular Formula C20H18N6Na2O9S
Quantity 1 g
Solubility Information Solubility aq. soln.: soluble.
InChI Key GRIXGZQULWMCLU-UHFFFAOYNA-L
SMILES [Na+].[Na+].COC1(NC(=O)C(C([O-])=O)C2=CC=C(O)C=C2)C2OCC(CSC3=NN=NN3C)=C(N2C1=O)C([O-])=O
IUPAC Name disodium 7-[2-carboxylato-2-(4-hydroxyphenyl)acetamido]-7-methoxy-3-{[(1-methyl-1H-1,2,3,4-tetrazol-5-yl)sulfanyl]methyl}-8-oxo-5-oxa-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Molecular Weight (g/mol) 564.44
Formula Weight 564.44
Chemical Name or Material Moxalactam sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.