missing translation for 'onlineSavingsMsg'
Learn More

MP Alumina A, Activity I, Acid pH, MP Biomedicals™

Product Code. 17845616 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 kg
Unit Size:
50kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17845616 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17845616 Supplier MP Biomedicals Supplier No. 0202159
  • ONLINE EXCLUSIVE PRICE
    2765.00€
    2630.25€ / 50kg
    Save 134.75€
    4% Off
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MP EcoChrom™ Alumina A, Activity I are a good all-round adsorbents for laboratory use as well as for industrial use. It is used as an adsorbent, desiccating agent, and catalyst, and in the manufacture of dental cements and refractories.

MP EcoChrom™ Alumina A, Activity I are:

  • Good ''all-round'' adsorbents for laboratory.
  • Used as well as for industrial use.
  • They can be adjusted to lower activity by the addition of polar media, preferably water, but also higher alcohols, e.g. glycol or glycerol can be used.
  • An oxide of aluminum, occurring in nature as various minerals such as bauxite, corundum, etc.

It is used as:

  • An adsorbent
  • A desiccating agent
  • A catalyst
  • In the manufacture of dental cements and refractories
  • Free flowing powder

Application Notes:

  • MP EcoChrom™ Alumina A catalyses a variety of reactions that are useful industrially.
  • It is widely used to remove water from gas streams.
  • Also used for its hardness and strength.
  • It is widely used as an abrasive, including as a much less expensive substitute for industrial diamond.
  • Many types of sandpaper use aluminium oxide crystals.
  • Acidic alumina, is the weakest and least often used type.
  • Alumina is useful for separation of acid pigments and strong acids.

It can cause:

  • Polymerization
  • Condensation
  • Dehydration reactions

Notes:

  • Neither acetone nor ethyl acetate should be used as eluants, the latter being subject to saponification.
  • As well, it is used as a dosimeter for radiation protection and therapy applications for its optically stimulated luminescence properties.

Usage Statement:

  • Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Key Applications:

  • Chromatography
  • Adsorbants
  • TLC
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point Undetermined
Melting Point ∼2072°C
CAS 1344-28-1
Color Off White
pH 4.5
Physical Form Powder
Quantity 50 kg
Molecular Formula Al2O3
MDL Number MFCD00003424
Synonym Alumina, Aluminum oxide, Activated aluminum oxide
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES [O--].[O--].[O--].[Al+3].[Al+3]
IUPAC Name dialuminium(3+) trioxidandiide
Molecular Weight (g/mol) 101.96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.