missing translation for 'onlineSavingsMsg'
Learn More

MP Alumina R for Isotope, Acid pH, 63 to 200 μm, MP Biomedicals™

Product Code. 17885616 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 kg
Unit Size:
50kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17885616 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17885616 Supplier MP Biomedicals Supplier No. 0206031
  • ONLINE EXCLUSIVE PRICE
    3630.00€
    3453.45€ / 50kg
    Save 176.55€
    4% Off
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MP EcoChrom™ Alumin R is an acid alumina, developed for use in isotope chemistry.

Application Notes:

  • MP EcoChrom™ Alumina R catalyses a variety of reactions that are useful industrially.

It is used:

  • To remove water from gas streams for its hardness and strength.
  • As an abrasive, including as a much less expensive substitute for industrial diamond.
  • Many types of sandpaper use aluminium oxide crystals.
  • In chemistry laboratories, alumina is a medium for chromatography, available in basic (pH 9.5), acidic (pH 4.5 when in water) and neutral formulations.
  • Acidic alumina, is the weakest and least often used type.
  • Alumina is useful for separation of acid pigments and strong acids.

It can cause:

  • Polymerization condensation dehydration reactions.
  • Neither acetone nor ethyl acetate should be used as eluants, the latter being subject to saponification.
  • As well, it is used as a dosimeter for radiation protection and therapy applications for its optically stimulated luminescence properties.

Usage Statement:

  • Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Key Applications:

  • Chromatography
  • Adsorbants
  • TLC
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point ∼2980°C
Melting Point ∼2072°C
CAS 1344-28-1
Color White
pH 4.3 ± 0.1
Physical Form Powder
Quantity 50 kg
Molecular Formula Al2O3
MDL Number MFCD00003424
Synonym Alumina, Aluminum oxide, Activated aluminum oxide
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES [O--].[O--].[O--].[Al+3].[Al+3]
IUPAC Name dialuminium(3+) trioxidandiide
Molecular Weight (g/mol) 101.96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.