missing translation for 'onlineSavingsMsg'
Learn More

MP EcoChrome™ Alumina A, Activity I, 63–200μm, MP Biomedicals™

Product Code. 11313529 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11313529 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11313529 Supplier MP Biomedicals Supplier No. 0202102
  • ONLINE EXCLUSIVE PRICE
    82.70€
    78.75€ / 1kg
    Save 3.95€
    4% Off
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used in laboratories (e.g. chromatography medium) and for industrial uses as a good all-around adsorbent, desiccating agent and catalyst. MP EcoChrome™ Alumina A, Activity I, 63–200μm, MP Biomedicals can be adjusted to lower activity by t

Alumina A is an oxide of aluminum, occurring in nature as various minerals such as bauxite, corundum, etc. It can be adjusted to lower activity by the addition of polar media, preferably water, but also higher alcohols, e.g. glycol or glycerol can be used. It catalyzes a variety of reactions that are useful industrially.

  • In chemistry laboratories, as a medium for chromatography (available in basic, acidic and neutral formulations), as well as for alcohols, hydrocarbons, steroids, alkaloids and natural pigments
  • As an abrasive, due to its hardness and strength (e.g. as an inexpensive substitute for industrial diamond; used in many types of sandpaper in crystal form)
  • Widely used to remove water from gas streams
  • In the manufacture of dental cements and refractories
  • Neutral alumina, (less active than the basic form) is useful for separation of aldehydes, ketones, quinones, esters, lactones, and glucosides
  • Acidic alumina (pH 4.5 when in water), is the weakest and least often used type
  • Alumina is useful for separation of acid pigments and strong acids
  • It can cause polymerization, condensation and dehydration reactions
  • Neither acetone nor ethyl acetate should be used as eluants, the latter being subject to saponification
  • As well, it is used as a dosimeter for radiation protection and therapy applications for its optically stimulated luminescence properties

Specifications

CAS 1344-28-1
Physical Form Powder
Quantity 1 kg
Molecular Formula Al2O3
MDL Number MFCD00003424
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
Solubility Information Insoluble in water
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES [O--].[O--].[O--].[Al+3].[Al+3]
IUPAC Name dialuminum;oxygen(2-)
Molecular Weight (g/mol) 101.96
PubChem CID 9989226
Formula Weight 101.96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.