missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPP7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12983-1-AP-150UL
This item is not returnable.
View return policy
Description
Acts as an important adapter that promotes epithelial cell polarity and tight junction formation via its interaction with DLG1. Involved in the assembly of protein complexes at sites of cell-cell contact.Specifications
| MPP7 | |
| Polyclonal | |
| Unconjugated | |
| MPP7 | |
| MAGUK p55 subfamily member 7, MPP7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 143098, 307035, 75739 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5T2T1, Q5U2Y3, Q8BVD5 | |
| Mpp7 | |
| MPP7 Fusion Protein Ag3612 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction